Learn More
Sino Biological Recombinant Cynomolgus, Rhesus PEAR1 Protein (ECD, His Tag) 100 µg
Catalog No. 5000656687
Encompass_Preferred
A DNA sequence encoding the Cynomolgus / Rhesus PEAR1 (XP_073896467.1) (Leu21-Leu755) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-56687 Supplier Sino Biological Supplier No. 91100C08C100
A DNA sequence encoding the Cynomolgus / Rhesus PEAR1 (XP_073896467.1) (Leu21-Leu755) was expressed with a polyhistidine tag at the C-terminus.