Learn More
Sino Biological Recombinant Cynomolgus Semaphorin 4A Protein (ECD, His Tag) 100 µg
Supplier:  Sino Biological 91107C08H100
Encompass_Preferred
A DNA sequence encoding the Cynomolgus Semaphorin 4A (G7NV79) (Met1-His683) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-006-56837