Learn More
BioVendor, LLC PROTEIN Cystatin C Human E. coli, 0.1 MG
SDP
Supplier:  BioVendor, LLC RD172009100H
Encompass
Type Recombinant protein Description Total 129 AA. Mw: 14.5 kDa (calculated). UniProtKB acc.no. P01034. N-terminal His-tag, 9 extra AA (highlighted). Amino Acid Sequence MKHHHHHHASSPGKPPRLVGGPMDASVEEEGVRRALDFAVGEYNKASNDMYHSRALQVVRARKQIVAGVNYFLDVELGRTTCTKTQPNLDNCPFHDQPHLKRKAFCSFQIYAVPWQGTMTLSKSTCQDA Source E. coli Purity Purity as determined by densitometric image analysis: >95% Endotoxin < 0.1 EU/μg Applications Western blotting, ELISA
Catalog No. 50-894-298