Learn More
Creative Biomart Pineapple stem bromelain
Supplier:  Creative Biomart THP0153
Encompass_Preferred
Bromelain is a category of protein-digesting enzymes that are obtained from the fruit or stem of pineapples. Although both fruit bromelain are prepared differently and contain different enzymatic compositions. Bromelain is consequently a composite mixture of several different endopeptidases that can facilitate many different reactions with many different substrates.
Catalog No. 50-208-4204