Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein Disulfide Isomerase from bovine liver >95% (SDS-PAGE), >=100 units/mg protein, lyophilized powder | Purity: >95% (SDS-PAGE) | 37318-49-3 | MFCD00163364 | 250UG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences P3818250UG
Encompass
Protein Disulfide Isomerase from bovine liver >95% (SDS-PAGE), >=100 units/mg protein, lyophilized powder | Purity: >95% (SDS-PAGE) | 37318-49-3 | MFCD00163364 | 250UG
Catalog No. 50-176-5083