Learn More
MOLECULAR DEPOT LLC Protein DNA Crystal - 48/1 mL
Catalog No. 5000013497
Encompass_Preferred
Crystallization Screen for Protein DNA Complexes is a crystallization screen consisting of 48 unique crystallization reagents that were proven to be successful in crystallizing protein-DNA complexes These 48 reagents were formulated after analyzing over 7000 published protein-DNA crystallization conditions and formulating unique reagents that included critical components for the growth of protein-DNA crystals such as the presence of polyamines PEG of various molecular weights divalent ions various salts reducing agents and other additives For in vitro research use only
Catalog No. 50-000-13497 Supplier MOLECULAR DEPOT LLC Supplier No. K1010003
Crystallization Screen for Protein DNA Complexes is a crystallization screen consisting of 48 unique crystallization reagents that were proven to be successful in crystallizing protein-DNA complexes These 48 reagents were formulated after analyzing over 7000 published protein-DNA crystallization conditions and formulating unique reagents that included critical components for the growth of protein-DNA crystals such as the presence of polyamines PEG of various molecular weights divalent ions various salts reducing agents and other additives For in vitro research use only