Learn More
Medchemexpress LLC Protein E7 HPV16 H 5ug
SDP
Catalog No. 5000182713
Encompass_Preferred
HPV16 E7 is a viral phosphoprotein with oncogenic activity belonging to the human papillomavirus early protein family HPV16 E7 activates the cell cycle by releasing E2F through proteasome degradation of pRB HPV16 E7 can cooperate with E6 to immortalize cells and neutralize the toxicity of E5 while activating the AP-1/NF- B pathway and downregulating E-cadherin to enhance migration and invasion making it a core target for cervical cancer Protein E7 HPV (His) is a recombinant HPV E7 protein expressed from E coli with an N-6 His tag
Catalog No. 50-001-82713 Supplier Medchemexpress LLC Supplier No. HYP722595UG
HPV16 E7 is a viral phosphoprotein with oncogenic activity belonging to the human papillomavirus early protein family HPV16 E7 activates the cell cycle by releasing E2F through proteasome degradation of pRB HPV16 E7 can cooperate with E6 to immortalize cells and neutralize the toxicity of E5 while activating the AP-1/NF- B pathway and downregulating E-cadherin to enhance migration and invasion making it a core target for cervical cancer Protein E7 HPV (His) is a recombinant HPV E7 protein expressed from E coli with an N-6 His tag