Learn More
Sino Biological Protein EBOV EBOV-NP
Catalog No. 5000131060
Encompass_Preferred
A DNA sequence encoding the Bundibugyo ebolavirus(Subtype Sudan strain Gulu) NP (YP-138520 1) (Gly630-Asp738) was expressed with an N-terminal polyhistidine tagged AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-001-31060 Supplier Sino Biological Supplier No. 40444V44E1B20
A DNA sequence encoding the Bundibugyo ebolavirus(Subtype Sudan strain Gulu) NP (YP-138520 1) (Gly630-Asp738) was expressed with an N-terminal polyhistidine tagged AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed