Learn More
Sino Biological Protein EBV-gp42 Biotin 20UG
Catalog No. 5001073665
Encompass_Preferred
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) EBV gp42/glycoprotein 42 (P03205) (Gly34-Ser223) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73665 Supplier Sino Biological Supplier No. 41200V49HB20
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) EBV gp42/glycoprotein 42 (P03205) (Gly34-Ser223) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed