Learn More
Key Scientific Products KPEX, Bacterial Protein Extraction Reagent, 10mL
SDP
Catalog No. NC0799802
Encompass_Preferred
A protein extraction-lysis buffer for use with Rosco antimicrobial resistance mechanism detection kits.
Catalog No. NC0799802 Supplier Key Scientific Products Supplier No. K2380
A protein extraction-lysis buffer for use with Rosco antimicrobial resistance mechanism detection kits.