Learn More
Biochain Institute Inc Total Protein Extraction Kit: TM Buffer, 13 ml/PK
SDP
Supplier:  Biochain Institute Inc K30110101
Encompass_Preferred
BioChain's Total Protein Extraction Kit is designed for rapid and efficient isolation of total protein lysates from virtually any tissue or cell sample. The system utilizes our proprietary buffer that has been optimized to prepare the highest quality total protein extractions. The kit also includes our proprietary protease inhibitor cocktail to prevent protein degradation. In conjunction, these two components result in protein extracts that are native and may be used for downstream applications such as SDS-PAGE, isoelectric focusing and 2-D gel electrophoresis, Western blotting, tissue-specific protein expression analysis, immunoprecipitation, protein-protein interaction studies, gel mobility shift analysis, DNA-protein interaction studies, antibody production, and enzymatic activity assays. Our Total Protein Extraction Kit is user-friendly and contains enough reagents for isolating total proteins from up to 5 grams of tissue or 125 million cultured cells. The kit is also scalable.
Catalog No. 50-183-015