Learn More
Biochain Institute Inc CNM Compartmental Protein Extraction Kit, 1 kit/PK
SDP
Catalog No. 50183004
Encompass_Preferred
BioChain's compartmental protein extraction kit is provided with protease inhibitor cocktail to prevent protein degradation. One kit contains four reagents enough to enrich three compartment proteins from 5 grams tissues or cells. The efficiency of subcellular fractionation has been investigated by SDS-PAGE and immunoblotting of selected marker proteins.
Catalog No. 50-183-004 Supplier Biochain Institute Inc Supplier No. K3012010
BioChain's compartmental protein extraction kit is provided with protease inhibitor cocktail to prevent protein degradation. One kit contains four reagents enough to enrich three compartment proteins from 5 grams tissues or cells. The efficiency of subcellular fractionation has been investigated by SDS-PAGE and immunoblotting of selected marker proteins.