Learn More
Biochain Institute Inc Membrane Protein Extraction Kit: Buffer C, 22 ml/PK
SDP
Catalog No. 50183012
Encompass_Preferred
BioChain's Membrane Protein Extraction Kit is designed to quickly isolate membrane proteins from mammalian tissues and cells. It is provided with protease inhibitor cocktail to prevent protein degradation. One kit contains four reagents enough to enrich three compartment proteins from 5 grams tissues or about 125 million cells. The efficiency of subcellular fractionation has been investigated by SDS-PAGE and immunoblotting of selected marker proteins.
Catalog No. 50-183-012 Supplier Biochain Institute Inc Supplier No. K30140051
BioChain's Membrane Protein Extraction Kit is designed to quickly isolate membrane proteins from mammalian tissues and cells. It is provided with protease inhibitor cocktail to prevent protein degradation. One kit contains four reagents enough to enrich three compartment proteins from 5 grams tissues or about 125 million cells. The efficiency of subcellular fractionation has been investigated by SDS-PAGE and immunoblotting of selected marker proteins.