Learn More
Covaris Inc truXTRAC Protein Extraction Buffer DF
SDP
Catalog No. NC3496236
Encompass
Efficient detergent-free protein extraction compatible with multiple downstream analysis methods. Stringent urea-based reagent includes optional additives EDTA, protease inhibitors and NaCl to allow optimization for your experimental needs. A superior alternative to detergent containing reagents for total protein extraction.
Catalog No. NC3496236 Supplier Covaris Inc Supplier No. 520093
Efficient detergent-free protein extraction compatible with multiple downstream analysis methods. Stringent urea-based reagent includes optional additives EDTA, protease inhibitors and NaCl to allow optimization for your experimental needs. A superior alternative to detergent containing reagents for total protein extraction.