Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein Extraction Reagent
Catalog No. 502905154
Encompass
Protein Extraction Reagent Type 4 is a highly premixed prequalified and effective protein solubilization and extraction reagent composed of urea thiourea Trizma(R) and detergent C7BzO (C0856).
Catalog No. 50-290-5154 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C03564BTL
Protein Extraction Reagent Type 4 is a highly premixed prequalified and effective protein solubilization and extraction reagent composed of urea thiourea Trizma(R) and detergent C7BzO (C0856).