Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein G Agarose
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11719416001
Encompass
Protein G was initially isolated from G148 a human group G Streptococcal strain. It is a cell wall protein and shows high affinity to IgG (immunoglobulin G). It has a putative molecular weight of 30 000Da. It interacts with all subclasses of human IgG and also binds to mouse rabbit and goat IgG.
Catalog No. 50-290-5471