Learn More
Ubpbio PROTEIN G AGAROSE, 5ML
Catalog No. 501126964
Encompass_Preferred
Protein Purification Reagents – Protein G Resin (UBPBio catalog# P5040-1, P5040-5, P5040-25) – Protein G XPure Agarose Resin consists of recombinant protein G covalently bound to crosslinked agarose beads, which minimizes leakage of Protein A/G and allow reuse the resin. The protein G resin has a binding capacity of ~ 20 mg human IgG/ml resin and works well in batch or FPLC column purifications.
Catalog No. 50-112-6964 Supplier Ubpbio Supplier No. P50405
Protein Purification Reagents – Protein G Resin (UBPBio catalog# P5040-1, P5040-5, P5040-25) – Protein G XPure Agarose Resin consists of recombinant protein G covalently bound to crosslinked agarose beads, which minimizes leakage of Protein A/G and allow reuse the resin. The protein G resin has a binding capacity of ~ 20 mg human IgG/ml resin and works well in batch or FPLC column purifications.