Learn More
Cell Signaling Technology Protein G Agarose Beads, Immunoprecipitation, 200 μL
SDP
Catalog No. 502050793
Encompass
These affinity matrix beads are designed for the small-scale isolation of immunocomplexes in immunoprecipitation assays. Protein G is covalently coupled to agarose beads, providing high affinity for various IgG subclasses from multiple species, including human, rabbit, mouse, rat, and sheep.
  • Features a bead diameter of 50-150 micron per bead.
  • Provides a binding capacity of 20 mg human IgG/mL.
  • Supplied as a 50% slurry in 20% ethanol solution.
  • Stable for 36 months when stored at 4°C.
  • Validated for use in immunoprecipitation applications.
Catalog No. 50-205-0793 Supplier Cell Signaling Technology Supplier No. 37478P
These affinity matrix beads are designed for the small-scale isolation of immunocomplexes in immunoprecipitation assays. Protein G is covalently coupled to agarose beads, providing high affinity for various IgG subclasses from multiple species, including human, rabbit, mouse, rat, and sheep.
  • Features a bead diameter of 50-150 micron per bead.
  • Provides a binding capacity of 20 mg human IgG/mL.
  • Supplied as a 50% slurry in 20% ethanol solution.
  • Stable for 36 months when stored at 4°C.
  • Validated for use in immunoprecipitation applications.