Learn More
Cell Signaling Technology Protein G Agarose Beads, Immunoprecipitation, 1 mL
SDP
Catalog No. NC1382746
Encompass
Protein G Agarose Beads are an affinity matrix designed for the small-scale isolation of immunocomplexes in immunoprecipitation (IP) assays. Protein G is covalently coupled to agarose beads and exhibits high affinity for various subclasses of IgG from many species, including human, rabbit, mouse, rat, and sheep. The beads are supplied as a 50% slurry in a 20% ethanol solution and are suitable for research applications requiring the efficient capture of antibodies.
  • Bead diameter: ~50-150 micron
  • Binding capacity: ~20 mg human IgG/mL
  • Stable for 36 months when stored at 4°C
  • Validated for immunoprecipitation protocols
Catalog No. NC1382746 Supplier Cell Signaling Technology Supplier No. 37478S
Protein G Agarose Beads are an affinity matrix designed for the small-scale isolation of immunocomplexes in immunoprecipitation (IP) assays. Protein G is covalently coupled to agarose beads and exhibits high affinity for various subclasses of IgG from many species, including human, rabbit, mouse, rat, and sheep. The beads are supplied as a 50% slurry in a 20% ethanol solution and are suitable for research applications requiring the efficient capture of antibodies.
  • Bead diameter: ~50-150 micron
  • Binding capacity: ~20 mg human IgG/mL
  • Stable for 36 months when stored at 4°C
  • Validated for immunoprecipitation protocols