Learn More
Seracare Life Sciences Inc PROTEIN G AGAROSE PURIF KIT
Catalog No. 5067523
Encompass_Preferred
The Protein G Agarose Purification Kit contains all the reagents needed to purify immunoglobulin molecules from several mammalian species. The kit consists of recombinant Protein G immobilized onto 4% cross-linked agarose. Recombinant Protein G is modified to eliminate binding of albumin and cell surface binding as occurs with native protein G. The binding capacity is greater than 15 mg/mL of human IgG per ml of resin. Flexible kits provide resin that can be used in gravity flow columns or other automated systems. Columns can be simply regenerated leading to greater cost efficiency.
This item is not returnable. View return policy
Catalog No. 50-675-23 Supplier Seracare Life Sciences Inc Supplier No. 57200004
The Protein G Agarose Purification Kit contains all the reagents needed to purify immunoglobulin molecules from several mammalian species. The kit consists of recombinant Protein G immobilized onto 4% cross-linked agarose. Recombinant Protein G is modified to eliminate binding of albumin and cell surface binding as occurs with native protein G. The binding capacity is greater than 15 mg/mL of human IgG per ml of resin. Flexible kits provide resin that can be used in gravity flow columns or other automated systems. Columns can be simply regenerated leading to greater cost efficiency.