Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein G from Streptococc5MG
Catalog No. 502901168
Encompass
Genetically engineered truncated protein G retains affinity for IgG but lacks albumin- and fab- binding sites and membrane-binding regions.
Catalog No. 50-290-1168 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 080625MGF
Genetically engineered truncated protein G retains affinity for IgG but lacks albumin- and fab- binding sites and membrane-binding regions.