Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Protein G-HRP
SDP

Catalog No. OB740605 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
1.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB740605 1.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB740605 Supplier SouthernBiotech™ Supplier No. 740605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certifications

Horseradish Peroxidase; 1.0 mL

Specifications

Formulation Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added
For Use With (Application) ELISA
Content And Storage 2°C to 8°C
Quantity 1.0 mL
Form Liquid

Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.