Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein G Mag SepharoseR C
Catalog No. 502900072
Encompass
Protein G Mag Sepharose(R) are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation applications. Based on Sepharose(R) with Protein G as ligand. Protein G has high affinity for the fc region of IgG antibodies.
Catalog No. 50-290-0072 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28951379
Protein G Mag Sepharose(R) are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation applications. Based on Sepharose(R) with Protein G as ligand. Protein G has high affinity for the fc region of IgG antibodies.