Learn More
Active Motif ChIP-IT Protein G Magnetic Beads
SDP
Catalog No. NC1368134
Encompass
Active Motif's ChIP-IT Protein G Magnetic Beads are the same Protein G magnetic beads that are included in our line of ChIP-IT Express Kits. The Protein G Magnetic Beads provide low background enrichment and streamline wash steps, making it possible to perform ChIP in just 1 day. For more complete information, please visit the Active Motif website.
Catalog No. NC1368134 Supplier Active Motif Supplier No. 53033
Active Motif's ChIP-IT Protein G Magnetic Beads are the same Protein G magnetic beads that are included in our line of ChIP-IT Express Kits. The Protein G Magnetic Beads provide low background enrichment and streamline wash steps, making it possible to perform ChIP in just 1 day. For more complete information, please visit the Active Motif website.