Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Protein G Magnetic Beads, 1.5 μm, 5 mL
SDP

Catalog No. 502050795
Encompass

Protein G Magnetic Beads function as an affinity matrix for the small-scale isolation of immunocomplexes in immunoprecipitation assays. Protein G is covalently coupled to magnetic particles, enabling efficient separation from solution when used with magnetic separation racks. This method eliminates centrifugation steps, reduces sample loss, and improves washing efficiency.

  • Provides high affinity for IgG subclasses from various species including human, rabbit, mouse, rat, and sheep.
  • Features a bead diameter of ∼1.5 μm.
  • Offers a binding capacity of >0.2 μg rabbit IgG/μl bead slurry.
  • Supplied in PBS, 0.1% BSA, 0.004% EDTA, and 0.075% sodium azide.
  • Storage at 4°C is required; stable for 12 months.
  • Do not freeze, dry, or centrifuge the beads to avoid irreversible aggregation.

Catalog No. 50-205-0795 Supplier Cell Signaling Technology Supplier No. 70024L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein G Magnetic Beads function as an affinity matrix for the small-scale isolation of immunocomplexes in immunoprecipitation assays. Protein G is covalently coupled to magnetic particles, enabling efficient separation from solution when used with magnetic separation racks. This method eliminates centrifugation steps, reduces sample loss, and improves washing efficiency.

  • Provides high affinity for IgG subclasses from various species including human, rabbit, mouse, rat, and sheep.
  • Features a bead diameter of ∼1.5 μm.
  • Offers a binding capacity of >0.2 μg rabbit IgG/μl bead slurry.
  • Supplied in PBS, 0.1% BSA, 0.004% EDTA, and 0.075% sodium azide.
  • Storage at 4°C is required; stable for 12 months.
  • Do not freeze, dry, or centrifuge the beads to avoid irreversible aggregation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.