Learn More
Bioworld PROTEIN G (RECOMBINANT)
Supplier:  Bioworld 220606782
Encompass_Preferred
Recombinant Protein G Recombinant Protein G contains only two IgG binding domain, but lacks the albumin binding domain to ensure the maximum specific IgG binding capacity. Protein G binds to all IgG subclasses from human, mouse and rat ...
Catalog No. 50-253-777