Learn More
Genscript Corporation Protein G Resin
Catalog No. NC1466772
Encompass
GenScript Protein G Resin is an affinity chromatography medium designed for easy, one-step purification of classes, subclasses and fragments of immunoglobulins from biological fluids and cell culture media. The recombinant protein G ligand is coupled to 4% highly cross-linked agarose. The static binding capacity of Protein G Resin is over 20 mg sheep IgG/ml settled resin. The dynamic binding capacity will vary depending on several factors such as target antibody, flow rate etc.
Catalog No. NC1466772 Supplier Genscript Corporation Supplier No. L00209
GenScript Protein G Resin is an affinity chromatography medium designed for easy, one-step purification of classes, subclasses and fragments of immunoglobulins from biological fluids and cell culture media. The recombinant protein G ligand is coupled to 4% highly cross-linked agarose. The static binding capacity of Protein G Resin is over 20 mg sheep IgG/ml settled resin. The dynamic binding capacity will vary depending on several factors such as target antibody, flow rate etc.