Learn More
Genscript Corporation Protein G Resin FF
Catalog No. 5000057105
Encompass
GenScript Protein G Resin FF is an affinity chromatography medium designed for easy one-step purification of classes subclasses and fragments of immunoglobulins from biological fluids and from cell culture media
Catalog No. 50-000-57105 Supplier Genscript Corporation Supplier No. L0066425
GenScript Protein G Resin FF is an affinity chromatography medium designed for easy one-step purification of classes subclasses and fragments of immunoglobulins from biological fluids and from cell culture media