Learn More
Genscript Corporation Protein G Resin FF
Supplier: Genscript Corporation L00664100
GenScript Protein G Resin FF is an affinity chromatography medium designed for easy one-step purification of classes subclasses and fragments of immunoglobulins from biological fluids and from cell culture media