Learn More
Bioworld PROTEIN G SEPAROPOREAGARO
Catalog No. 50197738
Encompass
PROTEIN G SEPAROPORE (AGAROSE) 4B: Protein-G Separopore 4B Affinity matrix with immobilized recombinant protein-G (from Streptococcus sp.) by CNBr activation coupling method.; For purification and/or fractionation of IgG subclasses which...
Catalog No. 50-197-738 Supplier Bioworld Supplier No. 201810302
PROTEIN G SEPAROPORE (AGAROSE) 4B: Protein-G Separopore 4B Affinity matrix with immobilized recombinant protein-G (from Streptococcus sp.) by CNBr activation coupling method.; For purification and/or fractionation of IgG subclasses which...