Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein G SepharoseR 4 Fas
Catalog No. 502908657
Encompass
Protein G Sepharose(R) 4 Fast Flow is recombinant protein G coupled to Sepharose(R) 4 Fast Flow.Protein G Sepharose(R) 4 Fast Flow has recombinant protein G immobilized by the cyanogen bromide (CNBr) method to Sepharose(R) 4 Fast Flow. Protein G exhibit binding specificities that complement Protein A media and binds to the Fc region of IgG from a variety of mammalian species. Protein G Sepharose(R) 4 Fast Flow may be used to isolate and purify classes subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.As member of the BioProcess media range Protein G Sepharose(R) 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.
Catalog No. 50-290-8657 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17061805
Add to Cart
Add to Cart
Request a Quote
Protein G Sepharose(R) 4 Fast Flow is recombinant protein G coupled to Sepharose(R) 4 Fast Flow.Protein G Sepharose(R) 4 Fast Flow has recombinant protein G immobilized by the cyanogen bromide (CNBr) method to Sepharose(R) 4 Fast Flow. Protein G exhibit binding specificities that complement Protein A media and binds to the Fc region of IgG from a variety of mammalian species. Protein G Sepharose(R) 4 Fast Flow may be used to isolate and purify classes subclasses and fragments of immunoglobulins from any biological fluid or cell culture medium.As member of the BioProcess media range Protein G Sepharose(R) 4 Fast Flow meets industrial demands with security of supply and comprehensive technical and regulatory support.