Learn More
Sigma Aldrich Fine Chemicals Biosciences Protein G SepharoseR fast
Catalog No. 502904564
Encompass
Protein G is an immunoglobulin (IgG)-binding bacterial cell wall protein isolated from group G streptococcal strain G-148. This protein can be extracted from the cells by papain digestion and purified by the sequential use of ion-exchange chromatography on DEAE-Sephadex affinity chromatography on Sepharose-coupled human IgG and gel chromatography on Sephadex G-200. The binding between protein G and various polyclonal and monoclonal IgG is basically pH dependent between 2.8 and 10 with the strongest binding at pH 4 and 5 and weakest at pH 10. It acts as a powerful reagent for the detection of IgG.P3296-5Ml updated product number is GE17-0618-01
Catalog No. 50-290-4564 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P32961ML
Protein G is an immunoglobulin (IgG)-binding bacterial cell wall protein isolated from group G streptococcal strain G-148. This protein can be extracted from the cells by papain digestion and purified by the sequential use of ion-exchange chromatography on DEAE-Sephadex affinity chromatography on Sepharose-coupled human IgG and gel chromatography on Sephadex G-200. The binding between protein G and various polyclonal and monoclonal IgG is basically pH dependent between 2.8 and 10 with the strongest binding at pH 4 and 5 and weakest at pH 10. It acts as a powerful reagent for the detection of IgG.P3296-5Ml updated product number is GE17-0618-01