Learn More
Medchemexpress LLC Protein G (Staphylococcus aureus), recombinant, C-terminal His tag | 250UG
SDP
Catalog No. 5000177193
Encompass_Preferred
Protein G (C-His) is a recombinant Staphylococcus aureus-derived Protein G expressed in Escherichia coli with a C-terminal 6xHis tag. It is intended for use in binding IgG Fc regions for antibody purification and detection workflows.
  • Recombinant Staphylococcus aureus source
  • C-terminal 6xHis tag for purification and detection
  • Expressed in Escherichia coli expression system
  • High affinity binding to IgG Fc regions for purification and detection
  • Suitable for biochemical assays and immobilization
  • Provided in a 250 μg format
Catalog No. 50-001-77193 Supplier Medchemexpress LLC Supplier No. HYP700321250UG
Protein G (C-His) is a recombinant Staphylococcus aureus-derived Protein G expressed in Escherichia coli with a C-terminal 6xHis tag. It is intended for use in binding IgG Fc regions for antibody purification and detection workflows.
  • Recombinant Staphylococcus aureus source
  • C-terminal 6xHis tag for purification and detection
  • Expressed in Escherichia coli expression system
  • High affinity binding to IgG Fc regions for purification and detection
  • Suitable for biochemical assays and immobilization
  • Provided in a 250 μg format