Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Protein G-Unlabeled
SDP

Catalog No. OB740601L Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB740601L 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB740601L Supplier SouthernBiotech™ Supplier No. 740601L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SouthernBiotech binding proteins are useful for immunoprecipitation, purification, and ELISA applications.

Specifications

Conjugate Unconjugated
Formulation Supplied lyophilized from distilled water; No BSA, preservatives or amine-containing buffer salts added
For Use With (Application) ELISA, IP, Purification
Content And Storage -20°C
Quantity 5 mg

For Research Use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.