Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Gel-Loading Dye for SDS-PAGE, 2X (Dark-Purple Liq./Electrophor.), Fisher BioReagents

Catalog No. p-27980 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Poly Bottle
Poly Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP637-1 1 mL Poly Tube
BP637-5 5 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP637-1
This dye is added to protein samples prior to electrophoresis on SDS–PAGE gels.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Protein Gel-Loading Dye for SDS-PAGE, 2X
Absorbance 0.15 to 0.35 AU at 252nm, 0.25 to 0.40 AU, 0.32 to 0.47 AU
CAS 56-81-5, 77-86-1, 151-21-3, 7647-01-0
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Sodium Dodecyl Sulfate (<5%), Glycerol (<12%), Tris (<1.0%), Proprietary Component (<2%), and Water (80%).
Packaging Poly Tube
Color Purple
Quantity 1 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.