Learn More
LAMDA BIOTECH INC Protein Gel SafeStain for 1L
Catalog No. NC3726406
Encompass_Preferred
A fluorescent protein staining reagent designed for detection of proteins in polyacrylamide gels. Features rapid staining protocol and sensitivity suitable for proteomic applications.
Catalog No. NC3726406 Supplier LAMDA BIOTECH INC Supplier No. G003
A fluorescent protein staining reagent designed for detection of proteins in polyacrylamide gels. Features rapid staining protocol and sensitivity suitable for proteomic applications.