Learn More
Sino Biological Protein Grin2b ET-FREE 100UG
Catalog No. 5001073918
Encompass_Preferred
A DNA sequence encoding the Human Grin2b (Q13224) (Arg27-Ser555) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73918 Supplier Sino Biological Supplier No. 13101H08HUE100
A DNA sequence encoding the Human Grin2b (Q13224) (Arg27-Ser555) was expressed with a C-terminal polyhistidine tag