Learn More
Sino Biological Protein Grin2b ET-FREE 1MG
Catalog No. 5001073986
Encompass_Preferred
A DNA sequence encoding the Mouse Grin2b (Q01097) (Arg27-Asp557) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73986 Supplier Sino Biological Supplier No. 5A7701M08HUE1
A DNA sequence encoding the Mouse Grin2b (Q01097) (Arg27-Asp557) was expressed with a C-terminal polyhistidine tag