Learn More
Sino Biological Protein Grin2b ET-FREE 1MG
Catalog No. 5001074062
Encompass_Preferred
A DNA sequence encoding the Human Grin2b (Q13224) (Arg27-Ser555) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-74062 Supplier Sino Biological Supplier No. 13101H08HUE1
A DNA sequence encoding the Human Grin2b (Q13224) (Arg27-Ser555) was expressed with a C-terminal polyhistidine tag