Learn More
Sino Biological Influenza A H3N2 (A/Alaska/144/2015) Neuraminidase / NA Protein (ECD, His & AVI Tag), Biotinylated 20 µg
Catalog No. 5000656672
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Alaska/144/2015) Neuraminidase / NA (ALH36899.1) (Ile30-Ile469) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-56672 Supplier Sino Biological Supplier No. 41115V49BB20
A DNA sequence encoding the Influenza A H3N2 (A/Alaska/144/2015) Neuraminidase / NA (ALH36899.1) (Ile30-Ile469) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.