Learn More
Sino Biological Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA-specific B cell probe Protein Trimer (Y108F, AVI & His Tag), Biotinylated (MALS-verified) 20 µg
Catalog No. 5000656522
Encompass_Preferred
A DNA sequence encoding the Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA-specific B cell probe Protein Trimer (EPI744956) (Met1-Val524) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-56522 Supplier Sino Biological Supplier No. 40104V27HB20
A DNA sequence encoding the Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA-specific B cell probe Protein Trimer (EPI744956) (Met1-Val524) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.