Learn More
Sino Biological Protein HHV-6 HHV6-U54 100UG
Catalog No. 5001073709
Encompass_Preferred
A DNA sequence encoding the Human B lymphotropic virus HHV-6A (strain Uganda-1102) U54 (P24434) (Met1-Pro458) was expressed with a C-terminal polyhistidine tag
Catalog No. 50-010-73709 Supplier Sino Biological Supplier No. 41164V08B100
A DNA sequence encoding the Human B lymphotropic virus HHV-6A (strain Uganda-1102) U54 (P24434) (Met1-Pro458) was expressed with a C-terminal polyhistidine tag