Learn More
Sino Biological Protein HSV2-gB Biotin 20UG
Catalog No. 5001073713
Encompass_Preferred
A DNA sequence encoding the HSV 2 (strain 333) Glycoprotein B (translated amino acids of P06763) (Glu98-Ala730) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73713 Supplier Sino Biological Supplier No. 40935V49HB20
A DNA sequence encoding the HSV 2 (strain 333) Glycoprotein B (translated amino acids of P06763) (Glu98-Ala730) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed