Learn More
Sino Biological Protein HSV2 gD Biotin 20UG
Catalog No. 5001073932
Encompass_Preferred
A DNA sequence encoding the HSV 2 (strain 333) gD (P03172) (Lys26-Thr310) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-73932 Supplier Sino Biological Supplier No. 40936V49HB20
A DNA sequence encoding the HSV 2 (strain 333) gD (P03172) (Lys26-Thr310) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed