Learn More
Sino Biological Protein HTNV-GP Biotin 20UG
Catalog No. 5001074023
Encompass_Preferred
A DNA sequence encoding the Hantaan virus (strain 76-118) Envelopment polyprotein/GP (P08668) (Ser649-Asn1105) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed
Catalog No. 50-010-74023 Supplier Sino Biological Supplier No. 41192V49BB20
A DNA sequence encoding the Hantaan virus (strain 76-118) Envelopment polyprotein/GP (P08668) (Ser649-Asn1105) was expressed with a C-terminal polyhistidine tag followed by an AVI tag The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed