Learn More
BioVendor, LLC PROTEIN HUMAN ADIPONECTIN HEK293
SDP
Supplier:  BioVendor, LLC RD172023100
Encompass
TypeRecombinant proteinDescriptionTotal 234 AA. MW 25.5 kDa (calculated). UniProtKB acc.no. Q15848 (Glu19 – Asn244). C-Terminal tag (8 extra AA).Amino Acid SequenceETTTQGPGVLLPLPKGACTGWMAGIPGHPGHNGAPGRDGRDGTPGEKGEKGDPGLIGPKGDIGETGVPGAEGPRGFPGIQGRKGEPGEGAYVYRSAFSVGLETYVTIPNMPIRFTKIFYNQQNHYDGSTGKFHCNIPGLYYFAYHITVYMKDVKVSLFKKDKAMLFTYDQYQENNVDQASGSVLLHLEVGDQVWLQVYGEGERNGLYADNDNDSTFTGFLLYHDTNDYKDDDDKGlu 1 to Gln 5 were confirmed by N-terminal sequencingSourceHEK293PurityPurity : > 95% Biological ActivityThe biological activity of Adiponectin is measured by its ability to inhibit proliferation of mouse 3T3 fibroblasts stimulated by PDGF. The ED50 for the effect is typically 5-15 μg/mL.Endotoxin< 0.1 EU/ugApplicationsWestern blotting, ELISA, Cell culture and/or animal studies
Catalog No. 50-871-240