Learn More
Sino Biological Recombinant Human Alpha-fetoprotein Protein (His & AVI Tag), Avi-tag Biotinylated, HPLC-verified 20 µg
Catalog No. 5000656573
Encompass_Preferred
A DNA sequence encoding the Human Alpha-fetoprotein (P02771) (Met1-Val609) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-006-56573 Supplier Sino Biological Supplier No. 12177H49HB20
A DNA sequence encoding the Human Alpha-fetoprotein (P02771) (Met1-Val609) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.