Learn More
Sino Biological Protein Human ALPP ET-FREE 1MG
Catalog No. 5001074005
Encompass_Preferred
A DNA sequence encoding the Human ALPP (P05187-1) (Ile23-Asp506) was expressed with a polyhistidine tag at the C-terminus
Catalog No. 50-010-74005 Supplier Sino Biological Supplier No. 12387H08HUE1
A DNA sequence encoding the Human ALPP (P05187-1) (Ile23-Asp506) was expressed with a polyhistidine tag at the C-terminus