Learn More
Sino Biological Recombinant Human ATAD2 Protein (Bromodomain, His Tag), Active 10 µg
Catalog No. 5000656719
Encompass_Preferred
Recombinant human ATAD2A bromodomain (981-1108) was expressed in E. coli cells using an N-terminal His tag.
Catalog No. 50-006-56719 Supplier Sino Biological Supplier No. A22310H10
Add to Cart
Add to Cart
Recombinant human ATAD2A bromodomain (981-1108) was expressed in E. coli cells using an N-terminal His tag.