Learn More
Sino Biological Recombinant Human ATAD2 Protein (Bromodomain, His Tag), Active 50 µg
Catalog No. 5000656836
Encompass_Preferred
Recombinant human ATAD2A bromodomain (981-1108) was expressed in E. coli cells using an N-terminal His tag.
Catalog No. 50-006-56836 Supplier Sino Biological Supplier No. A22310H50
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human ATAD2A bromodomain (981-1108) was expressed in E. coli cells using an N-terminal His tag.